Cell culture plates and Multidishes, Nunclon™ Sphera™
The Nunclon™ Sphera™ culture surface allows cells to grow in suspension with virtually no cell attachment. The surface supports many different cell types and their ability to generate spheroids.
- Optimises spheroid formation
- Supports the consistent formation of cancer spheroids that simulate the 3D structures of tumour growth
- Promotes formation of embryoid bodies (EB)
The Nunclon™ Sphera™ surface minimises variability and supports the consistent formation of cancer spheroids that simulate the 3D structures of tumour growth, providing a better model system for studying tumour cell progression and efficacy of anticancer agents in vitro.
Nunclon™ Sphera™ surface demonstrates superior quality for embryoid body (EB) formation of pluripotent stem cells with minimal spontaneous differentiation. The resultant cells are able to differentiate into all three germ layers.
Frequently Bought Together