Acetic anhydride
Catalog # LACH20006CT0M1000
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
About this item
Synonyms: Acetyl oxide, Acetyl acetate
:
:102.09 g/mol
EINECS:203-564-8
UN:1715
REACH:01-2119486470-36
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:108-24-7
Formula:(CH₃CO)₂O
MDL Number:MFCD00008705
Boiling Pt:139.9
Melting Pt:–73 °C
Density:1.08 g/cm³ (20 °C)
Flash Pt:49
ADR:8 II
Merck Index:13,00057