Buffers, TRIS HCl solutions, pH range 7,2 to 8,5 (1 mol/L)
Supplier: Rockland Immunochemicals
Certificates
About this item
Tris is widely used as a component of buffer solutions, such as in TAE and TBE buffer, especially for solutions of nucleic acids.
These solutions were aseptically filtered through a Millipore 0,22 µm filter into clean, pre-sterilized containers. The products were tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Caution: These products ares for research use only and are not intended for therapeutic or diagnostic applications.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together