Single channel micro pipettes, variable volume, Acura® manual 825
Supplier: Socorex Isba
Certificates
About this item
Manual pipettes for all applications
- Single-handed volume setting
- Precision digital display visible at all time
- Justip™ adjustable tip ejector fitting most tips
Delivery: The Multisets include: Three Acura® 825 manual adjustable pipettes, Qualitips® samples, QC certificates, operating instructions
Frequently Bought Together