Order Entry
Switzerland
ContactUsLinkComponent
Anti-FGL2 Antibody Pair
Anti-FGL2 Antibody Pair
N° de catalogue ABNOH00010875-PW1
Fournisseur:  Abnova
Anti-FGL2 Antibody Pair
N° de catalogue ABNOH00010875-PW1
Fournisseur:  Abnova

Spécifications

  • Nom de l'antigène:
    Fibrinogen-like 2
  • Symbole de l'antigène:
    FGL2
  • Hôte:
    Multiple
  • Précipitation immunologique:
    Yes
  • Isotype:
  • Réactivité:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • ID de gène:
    10875
  • Synonymes antigène:
    T49|pT49
  • Température de stockage:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Taille:
    1 Set
  • Cdt:
    1 SET

Spécifications

À propos de cet article

This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-FGL2 (300 µg); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-FGL2 (50 µl)

Type:
Antigen: FGL2
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human