Anti-CK19 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The Cytokeratin 19 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Cytokeratin 19. This antibody reacts with human, mouse, rat. The Cytokeratin 19 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:500
Type: Primary
Antigen: CK19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- NOVUNB100-687SS
- NOVUNB100-687
- Antigen-Symbol:
- CK19
- CK19
- Antigen-Name:
- Cytokeratin 19
- Cytokeratin 19
- Antigen-Synonyme:
- type I cytoskeletal 19|CK19|CK-19|K1CS|keratin-19|40-kd|K19|KRT19|cytokeratin-19|type I|keratin|40-kDa keratin intermediate filament
- type I cytoskeletal 19|CK19|CK-19|K1CS|keratin-19|40-kd|K19|KRT19|cytokeratin-19|type I|keratin|40-kDa keratin intermediate filament
- Antikörper-Typ:
- Primär
- Primär
- Klonalität:
- Polyklonal
- Polyklonal
- Konjugation:
- Unconjugated
- Unconjugated
- Reaktivität:
- Human
- Human
- Wirt:
- Rabbit
- Rabbit
- Gen-ID:
- 3880
- 3880
- Western blot:
- Yes
- Yes
- ImmunFluoreszenz:
- Yes
- Yes
- Lagerungspuffer:
- PBS and 30% Glycerol
- Lagertemperatur:
- Cool
- Cool
- Transporttemperatur:
- Ice
- Ice
Specifications
Frequently Bought Together