Anti-PARP1 Rabbit Polyclonal Antibody
Supplier: Biorbyt
Certificates
About this item
Type: Primary
Antigen: PARP1 (poly (ADP-ribose) polymerase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Specifications
- Catalog No:
- BIRBORB126564-200
- BIRBORB126564-100
- Antigen-Symbol:
- PARP1
- PARP1
- Antigen-Name:
- Poly (ADP-ribose) polymerase 1
- Poly (ADP-ribose) polymerase 1
- Antikörper-Typ:
- Primär
- Primär
- Klonalität:
- Polyklonal
- Polyklonal
- Konjugation:
- Unconjugated
- Unconjugated
- Reaktivität:
- Human
- Human
- Wirt:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- Modification:
- cleaved-Gly215
- cleaved-Gly215
- Reinigung:
- purified by affinity chromatography
- purified by affinity chromatography
- Konzentration:
- 1 mg/ml
- 1 mg/ml
- Lagerungspuffer:
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Lagertemperatur:
- Cool
- Cool
- Transporttemperatur:
- Ice
- Ice
Specifications
Frequently Bought Together