ContactUsLinkComponent
CELLCYTE GEN 2 INVERTED 4 CHANNEL
CELLCYTE G2 Live Cell Imaging and Analysis Inside Your Incubator
# de catalogue 77529-432
Fournisseur:  DISCOVER ECHO INC.
New Product
CELLCYTE G2 Live Cell Imaging and Analysis Inside Your Incubator
# de catalogue 77529-432
Fournisseur:  DISCOVER ECHO INC.
New Product
Spécifications
  • Description:
    CELLCYTE G2 complete imaging system, including microscopy unit, 4-channel CCU
Spécifications
A propos de cet article
Live cell imaging inside your incubator with 5 MP sCMOS monochrome camera (2448×2048 pixels).
  • Inverted, Enhanced phase contrast, fluorescence
  • Control unit with software for device control and acquisition
  • Includes PC and 22" HD Monitor
Fluorescence options:
3 Channel DAPI | GFP | mCherry
4 Channel DAPI | GFP | RFP | CY5
Included Analysis Modules:
Confluence
Cell counting
Apoptosis
Cytotoxicity
Transfection efficiency
Reporter genes
Immune cell cluster formations
4-Channel Fluorescence DAPI - EX:395/25 EM:450/30
GFP - EX:483/18 EM:525/25
RFP - EX:555/20 EM:595/30
CY5 - EX:635/20 EM:695/60
Customers Who Bought This Also Bought