Filter Sets - LED Light Cubes for ECHO Revolve 4K Hybrid Fluorescence Microscope (Upright + Inverted)
# de catalogue 77529-426
Fournisseur: DISCOVER ECHO INC.
New Product
Spécifications
- Description:LED light cubes narrowband EGFP (EX: 480/20 EM: 510/20 DM: 495 LPXR ET)