Filter Sets - LED Light Cubes for ECHO Revolve 4K Hybrid Fluorescence Microscope (Upright + Inverted)
# de catalogue 77529-422
Fournisseur: DISCOVER ECHO INC.
New Product
Spécifications
- Description:LED light cubes BFP (EX:405/20 EM: 460/50 DM: 425 lp)