ContactUsLinkComponent
FILTER SET FLUORESCENCE YFP 6008
Filter Sets - LED Light Cubes for ECHO Revolve 4K Hybrid Fluorescence Microscope (Upright + Inverted)
# de catalogue 77529-418
Fournisseur:  DISCOVER ECHO INC.
New Product
Filter Sets - LED Light Cubes for ECHO Revolve 4K Hybrid Fluorescence Microscope (Upright + Inverted)
# de catalogue 77529-418
Fournisseur:  DISCOVER ECHO INC.
New Product
Spécifications
  • Description:
    LED light cubes YFP (EX: 500/20 EM: 535/30 DM: 515 lp)