ContactUsLinkComponent
FILTER SET FLUORESCENCE CFP 6007
Filter Sets - LED Light Cubes for ECHO Revolve 4K Hybrid Fluorescence Microscope (Upright + Inverted)
# de catalogue 77529-416
Fournisseur:  DISCOVER ECHO INC.
New Product
Filter Sets - LED Light Cubes for ECHO Revolve 4K Hybrid Fluorescence Microscope (Upright + Inverted)
# de catalogue 77529-416
Fournisseur:  DISCOVER ECHO INC.
New Product
Spécifications
  • Description:
    LED light cubes CFP (EX: 445/20 EM: 510/40 DM: 455 lp)