Filter Sets - LED Light Cubes for ECHO Revolve 4K Hybrid Fluorescence Microscope (Upright + Inverted)
# de catalogue 77529-414
Fournisseur: DISCOVER ECHO INC.
New Product
Spécifications
- Description:LED light cubes ALEXA/AF 594 (EX: 580/30 EM: 635/50 DM: 594)