Filter Sets - LED Light Cubes for ECHO Revolve 4K Hybrid Fluorescence Microscope (Upright + Inverted)
# de catalogue 77529-412
Fournisseur: DISCOVER ECHO INC.
New Product
Spécifications
- Description:LED light cubes CY5 (EX: 640/30 EM: 690/50 DM: 660)