ContactUsLinkComponent
FILTER SET FLUORESCENCE TXRED 6004
Filter Sets - LED Light Cubes for ECHO Revolve 4K Hybrid Fluorescence Microscope (Upright + Inverted)
# de catalogue 77529-410
Fournisseur:  DISCOVER ECHO INC.
New Product
Filter Sets - LED Light Cubes for ECHO Revolve 4K Hybrid Fluorescence Microscope (Upright + Inverted)
# de catalogue 77529-410
Fournisseur:  DISCOVER ECHO INC.
New Product
Spécifications
  • Description:
    LED light cubes TXRED (EX: 560/40 EM: 635/60 DM: 600)