Filter Sets - LED Light Cubes for ECHO Revolve 4K Hybrid Fluorescence Microscope (Upright + Inverted)
# de catalogue 77529-406
Fournisseur: DISCOVER ECHO INC.
New Product
Spécifications
- Description:LED light cubes FITC (EX: 470/40 EM: 525/50 DM: 495)