ContactUsLinkComponent
FILTER SET FLUORESCENCE FITC 6002
Filter Sets - LED Light Cubes for ECHO Revolve 4K Hybrid Fluorescence Microscope (Upright + Inverted)
# de catalogue 77529-406
Fournisseur:  DISCOVER ECHO INC.
New Product
Filter Sets - LED Light Cubes for ECHO Revolve 4K Hybrid Fluorescence Microscope (Upright + Inverted)
# de catalogue 77529-406
Fournisseur:  DISCOVER ECHO INC.
New Product
Spécifications
  • Description:
    LED light cubes FITC (EX: 470/40 EM: 525/50 DM: 495)