Filter Sets - LED Light Cubes for ECHO Revolve 4K Hybrid Fluorescence Microscope (Upright + Inverted)
# de catalogue 77529-404
Fournisseur: DISCOVER ECHO INC.
New Product
Spécifications
- Description:LED light cubes DAPI (EX: 385/30 EM: 450/50 DM: 425)