Stainless Steel Raised Wire Floors, Tecniplast®
Catalog # 77322-378
Supplier: Tecniplast Usa
New Product
Specifications
- Description:Raised floor for cage size C, stainless steel wire
- Cat. no.:77322-378
Specifications
About this item
Stainless steel design with 30 mm elevation separates animals from bedding and waste.
Not required for standard operation but recommended for post-surgical animals or for those on restrictive diets. Help prevent foot pad swelling. Convenient handle design for easy insertion and removal. Autoclavable.
Customers Who Bought This Also Bought