ContactUsLinkComponent
CAGE LID SS WIRE SIZE F
Stainless Steel Wire Cage Lids, Tecniplast®
# de catalogue 77322-426
Fournisseur:  Tecniplast Usa
New Product
CAGE LID SS WIRE SIZE F
Stainless Steel Wire Cage Lids, Tecniplast®
# de catalogue 77322-426
Fournisseur:  Tecniplast Usa
New Product
Spécifications
  • Description:
    Inner fitting lid with hinged divider for cage size F
  • For Use With:
    Rats and hamsters
Spécifications
A propos de cet article
Desgined to ensure a perfect fit and secure lock throughout the complete range of Tecniplast® cages.
Stainless steel drop-in hoppers are autoclavable. A central divider prevents the animal from crawling into the hoppper, which can be used with or without a raised wire floor.
Secure and easy to handle, these autoclavable stainless steel wire lids have cross-wire members to prevent bending. With the exception of flat lids 95057-874 and 95057-888, all lids feature a U-shaped hopper for easy access to food. Hoppers won’t entrap food or contaminants. Wire spacing is 7 mm for most sizes; 19 mm spacing for guinea pig cage lid 10713-340. Hopper divider folds down, allowing lids to be stacked for storage. Flat lids 95057-874, 95057-888, and 10713-345 are for use without a raised floor. Incorporate stopper guard for water bottle sipper tube.
Customers Who Bought This Also Bought