Reusable Filter Animal Cage Covers, Tecniplast®
# de catalogue 77322-420
Fournisseur: Tecniplast Usa
New Product
Spécifications
- Description:Covers for cage size E
Spécifications
A propos de cet article
Securely fit Tecniplast® plastic animal cages.
- Filter covers permit atmospheric gas passage with minimal buildup of temperature and humidity
- Covers are constructed of transparent polysulfone with a stainless steel filter retainer frame
- Covers fit over water bottle
Replacement filters are ready-to-use and presized to fit neatly inside the cover.
Customers Who Bought This Also Bought