ContactUsLinkComponent
RAISED FLOOR SSWIRE SIZE D
Stainless Steel Raised Wire Floors, Tecniplast®
Catalog # 77322-392
Supplier:  Tecniplast Usa
New Product
RAISED FLOOR SSWIRE SIZE D
Stainless Steel Raised Wire Floors, Tecniplast®
Catalog # 77322-392
Supplier:  Tecniplast Usa
New Product
Specifications
  • Description:
    Raised floor for cage size D, stainless steel wire
  • Cat. no.:
    77322-392
Specifications
About this item
Stainless steel design with 30 mm elevation separates animals from bedding and waste.
Not required for standard operation but recommended for post-surgical animals or for those on restrictive diets. Help prevent foot pad swelling. Convenient handle design for easy insertion and removal. Autoclavable.
Customers Who Bought This Also Bought