ContactUsLinkComponent
FILTER COVER REUSABLE B
Reusable Filter Animal Cage Covers, Tecniplast®
Catalog # 77322-368
Supplier:  Tecniplast Usa
New Product
FILTER COVER REUSABLE B
Reusable Filter Animal Cage Covers, Tecniplast®
Catalog # 77322-368
Supplier:  Tecniplast Usa
New Product
Specifications
  • Description:
    Covers for cage size B
  • Cat. no.:
    77322-368
Specifications
About this item
Securely fit Tecniplast® plastic animal cages.
  • Filter covers permit atmospheric gas passage with minimal buildup of temperature and humidity
  • Covers are constructed of transparent polysulfone with a stainless steel filter retainer frame
  • Covers fit over water bottle
Replacement filters are ready-to-use and presized to fit neatly inside the cover.
Customers Who Bought This Also Bought