Spécifications
- Description:Cricut Easypress® 2 machine
- Dimensions:12×10"
Spécifications
A propos de cet article
These large heat presses are ideal for jumbo-size iron-on vinyl projects.
- Professional heat-transfer results without the guesswork
- Advanced heat plate design with ceramic-coated surface
- Compatible with all Infusible Ink™ products and most major brands of iron-on material
- Precise temperature control up to 205 °C (400 °F)
- Easy to learn, simple to use
- Insulated safety base and auto-shutoff feature
- Lightweight, portable, easy to store
Now with adjustable heat up to 205 °C (400 °F), Cricut EasyPress 2 is a no-stress heat press. Combining the speed of a heat press and the convenience of an iron, Cricut EasyPress® 2 provides fast, effortless results that really last, even after repeated washes. Eliminate guesswork with rigorously tested time and temperature settings for every project, apply heat, and when the beep sounds, it's done. Helpful safety features include an insulated Safety Base and auto-shutoff feature. Cricut EasyPress® 2 is lightweight, portable, easy to store, and compatible with most major brands of heat-transfer vinyl and all Infusible Ink™ products. And of course, it’s a perfect complement to Cricut cutting machines.