Spécifications
- Description:Cricut EasyPress® 3 machine
- Dimensions:9×9"
Spécifications
A propos de cet article
Say hello to the perfect press for heat transfer projects.
- Ideal for sweatshirts, banners, blankets, and more
- Connects to Cricut Heat™ app via Bluetooth® connectiona
- Precise temperature control up to 205 °C (400 °F)
- Advanced heat plate design with ceramic-coated surface
- Compatible with major brands of iron-on (HTV), Infusible Ink™ and sublimation products
Cricut EasyPress® 3 brings together the speed of a heat press, the convenience of an iron, and the simplicity of the Cricut Heat app. Press without the guesswork, and get fast, easy transfers. Simply send ideal time and temperature settings through Bluetooth® connection via the Cricut Heat app. Follow on-screen, step-by-step prompts that guide you at every turn. The ceramic-coated heat plate reaches temperatures up to 205 °C (400 °F) of dry, edge-to-edge heat. Enjoy a worry-free experience with safety features like an insulated safety base and auto-off. It’s compact, easy to store, and compatible with major brands of iron-on (HTV), Infusible Ink and sublimation products. A perfect complement to Cricut cutting machines.