CRICUT SMART IRON-ON VINYL WHITE 3FT
# de catalogue 470354-250
Fournisseur: D&H
New Product
Customers Who Bought This Also Bought
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Fournisseur: MilliporeSigma
$768.73 to $3,075.66
n-Heptane ≥99% (n-heptane) [≥99.9% (saturated C7 hydrocarbons)], OmniSolv®, Supelco®
Fournisseur: MilliporeSigma
$971.75 to $3,463.97
Hexane (mixture of isomers) ≥98.5% ACS for HPLC, Supelco®
Fournisseur: MilliporeSigma
$649.44 to $2,496.86
Traceable® Certified Reference Material (CRM) pH Buffers, Control Company
Fournisseur: Argos Technologies
$62.31
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
$635.20 to $2,541.50
VeriSpec™ Sulfate - SO₄²⁻ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Fournisseur: Ricca Chemical
$230.13