Cricut Joy™ Cutting Machine
# de catalogue 470354-254
Fournisseur: D&H
New Product
Spécifications
- Description:Cricut Joy Cutting Machine
Spécifications
A propos de cet article
Ultra-compact smart cutting machine, perfect for Makerspaces and schools.
- Works with 50+ materials, including vinyl, iron-on, and cardstock
- Cuts, writes, draws, and foils intricate designs with precision
- Free, easy-to-learn Design Space app
- Bluetooth wireless technology
Create custom cards, simple decals, and labels with this ultra-compact smart cutting machine. You can make fun or functional projects in 15 minutes or less. No design experience required. Works with a free companion app.
Delivery: Includes: Cricut Joy machine; Premium Fine-Point Blade + Housing; Fine Point Pen; Standard Grip Mat (4.5×6.5"); Welcome card; USB cable; Power adapter; Free trial subscription to Cricut Access™ (for new subscribers); 50 free, pre-designed projects in the Design Space app; Materials for a practice cut.
Customers Who Bought This Also Bought