Spécifications
- Color:Raspbery
- Description:Cricut EasyPress® Mini machine
Spécifications
A propos de cet article
Compact size ideal for unusually small or unique heat transfer projects.
- Customize unusual projects like shoes and stuffed animals
- Ceramic-coated heat plate for dry, even, edge-to-edge heat
- 3 heat settings for every project
- Insulated safety base and auto-shutoff feature
- Precision tip for added control and accuracy
- Lightweight, portable, easy to store
- Effortlessly glides over uneven surfaces
Mini but mighty; bring on the unusual objects and projects with Cricut Easypress® Mini™. Now you can get high-impact heat-transfer results in a thoughtful, compact design. Whether it's a uniquely shaped object or an extra-small project, including shoes, stuffed animals, and more, you'll have what it takes to make a successful transfer. Easily curve with contours, get between buttons, and even squeeze into seams. Three heat settings are all you need for every heat transfer project, including Infusible Ink™ projects. An insulated base and an auto-off feature are safety features you'll love. Plus it's lightweight, portable, and easy to store. Works with major brands of iron-on (HTV), Infusible Ink™ and sublimation products. A perfect complement to any Cricut cutting machine or larger Cricut heat press.