ContactUsLinkComponent
PLATES STREAK GLASS 1X2X1/4 PK10
Streak Plates, Glass
# de catalogue 470353-570
New Product
PLATES STREAK GLASS 1X2X1/4 PK10
Streak Plates, Glass
# de catalogue 470353-570
New Product
Spécifications
  • Description:
    Streak plates, glass
  • L×W:
    2×1"
  • Cat. no.:
    470353-570
Spécifications
A propos de cet article
Clear glass for mineral testing.
  • ¹/₄" thick glass
  • Overall dimensions: 2×1"
Designed for use in hardness testing of rocks and minerals.
Packaging: Plates are sold as packs of 10.
Customers Who Bought This Also Bought