VWR® Cleanroom Comfort Headbands, Made in USA
# de catalogue 77257-034
Fournisseur: VWR International
New Product
Spécifications
- Clothing Size:S
- Garment application:Cleanroom
- Description:VWR® cleanroom comfort headband
- Fabric:Continuous filament fiber stretch nylon
Spécifications
A propos de cet article
Sweat absorbing, user-friendly, disposable, and hygienic solution.
- Premium soft comfort fit
- Continuous filament fiber stretch nylon
- Sweat absorbing
- Cleanroom compatible
- Disposable
- Made in USA
The cleanroom comfort headband is designed for comfort and intended for use under a bouffant hat in cleanroom environments. These headbands are tailor made to ensure user comfort, crafted from continuous filament fiber, stretch nylon for a soft and comfortable cleanroom garment. This product minimizes the risk of contamination and eliminates the need for employees to use external products during business operations.
Customers Who Bought This Also Bought