ContactUsLinkComponent
GOWNS ISO 25GSM M/L CS100
RightCare Disposable Isolation Gowns
# de catalogue 77148-068
New Product
GOWNS ISO 25GSM M/L CS100
RightCare Disposable Isolation Gowns
# de catalogue 77148-068
New Product
Spécifications
  • Clothing Size:
    M/L
  • Color:
    Yellow
  • Cat. no.:
    77148-068
Spécifications
A propos de cet article
The RightCare isolation gown protects critical areas of the body, including shoulders, chest, legs, and arms. The fluid-resistant coating on the gown keeps fluid away from clothing, maintaining clean clothes that do not get contaminated. The gown is comfortable and features an open back with ties at the neck and at the waist. Arms are fully protected with long sleeves and an elastic cuff at the wrist.
  • Lightweight isolation gown designed for reliable, comfortable protection
  • High-quality 25 gsm spunbonded polypropylene construction
  • Full coverage of shoulders, chest, legs, and arms
  • Nonsterile disposable gowns for single-use only
  • Open back with tie closures at the neck and waist
  • Elastic wrist cuffs to ensure arms are properly protected
  • Fluid-resistant coated material keeps fluid out and away from clothing
Lightweight isolation gown designed for reliable, comfortable protection without restrictions on body movement. The disposable gown can be used for basic care, standard isolation, or as a gown for visitors in a clinical setting. High-quality 25 gsm spunbonded polypropylene construction provides excellent protection against fluids, particulates, or other contaminants.
Gowns are designed for single-use only. Always dispose of the gown promptly after use. This nonsterile gown should not be used for invasive procedures or surgery, although it will effectively protect clothing from becoming contaminated or stained.
Customers Who Bought This Also Bought