ContactUsLinkComponent
FACE SHIELD FULL BX3
RightCare Full Face Protective Shields with Anti-Fog Coating and Elastic Head Strap
# de catalogue 77148-086
New Product
FACE SHIELD FULL BX3
RightCare Full Face Protective Shields with Anti-Fog Coating and Elastic Head Strap
# de catalogue 77148-086
New Product
Spécifications
  • Color:
    Clear
  • Size:
    One size
  • Cat. no.:
    77148-086
Spécifications
A propos de cet article
The RightCare full face protective shield is ideal for use in all types of environments, and provides a full 180 degree coverage from flying debris, fluid droplets, aerosols, and all types of splatters. It can safely withstand heat, impacts, and chemicals. The face shield provides extra protection against germs when combined with the use of a mask.
  • Clear, transparent, anti-fog shield
  • High-strength PET thermoplastic material
  • Safe to use in the laboratory and other general medical field uses
  • Lightweight durable construction
  • One size, due to the ergonomic design with the flexible band
  • Most safety glasses or goggles, along with face masks, can still be worn underneath the shield
  • Can be easily disinfected for reuse
Full face protection against flying debris, fluid droplets, aerosols, and all types of splatters. The clear, transparent, and anti-fog shield effectively protects eyes, nose, and mouth without distorting the field of view. High-strength PET thermoplastic material can withstand heat, impact, chemicals, or other hazardous splashes. Safe to use in the laboratory and other general medical field uses. Equipped with a soft sponge padding on the forehead area and an elastic band that easily straps over the head.
180 degree coverage make it ideal for use in all types of environments such as in a hospital, clinic, dentist's office, laboratory, woodshop, school, pharmacy, and even work. Anti-fog coating helps to provide maximum visibility at all times.
Customers Who Bought This Also Bought