ASSAY KIT USP25 INHIBITOR SCREENING
# de catalogue 77039-278
Fournisseur: BIORTUS USA
New Product
Customers Who Bought This Also Bought
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Fournisseur: VWR International
$109.33 to $364.87
Sodium hypochlorite 12.5% in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$99.85 to $385.36
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$194.14 to $694.12