ACYRLIC BASE ASSY GAS
# de catalogue 76836-932
Fournisseur: LUCIDITY SYSTEMS LLC
New Product
Customers Who Bought This Also Bought
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Fournisseur: Thermo Scientific
260,09 $ to 2 400,95 $
Sodium carbonate, anhydrous ≥99.5% ACS, VWR Chemicals BDH®
Fournisseur: VWR International
129,84 $ to 1 081,96 $
Sulfuric acid 1.0 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
170,25 $ to 676,42 $
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
458,88 $ to 1 237,78 $
Sulfuric acid 0.1 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
67,74 $ to 772,55 $
Hydrochloric acid 10% (v/v) (1+9), VWR Chemicals BDH®
Fournisseur: VWR International
85,42 $ to 984,68 $