Nichitip Pipette Tips, Universal
# de catalogue 76808-744
Fournisseur: Nichiryo America
Spécifications
- Volume:100 - 1250 µl
- Sterile:+
- Tip Style:Low retention, extended length
Spécifications
A propos de cet article
These tips are made of 100% virgin polypropyleylene.
- High quality
- Free of detectable DNase, RNase, DNA
- Autoclavable
- Low retention
Made in the highest quality moulds, these tips can be used whenever the work demands a higher standard.
Customers Who Bought This Also Bought