VIAL PES .2UM LOEVAP FILT GRY SC CS100
Catalog # 76808-642
Supplier: THOMSON INSTRUMENT CO
Customers Who Bought This Also Bought
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Supplier: VWR International
$86.08 to $717.60
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
$118.61 to $1,128.11
SureSTART™ Caps for 2 ml Glass Snap Top Vials, Level 2 High-Throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
$45.12 to $48.54
WHEATON® LVI™ Large-Opening Screw-Thread Vials, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
$456.33
WHEATON® ABC Vial™ Large-Opening Screw-Thread Vials, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
$127.02
WHEATON® Battery Powered Crimpers and Decappers, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
$2,030.31