Spécifications
- Pore size:0.2 µm
- Color:Green
- Description:eXtreme/FV® Filter vial
- Cat. no.:76808-666
- Membrane:PTFE, tiered filtration
Spécifications
A propos de cet article
Thomson's filtration products are engineered and optimized to increase the efficiency of sample preparation without compromising the results.
Thomson eXtreme/FV® Filter Vials offer tiered filtration, and are designed to speed up sample prep and analysis on particulate laden samples. Save on costly repairs and service issues while eliminating multiple syringe filters, syringes, HPLC vials, and HPLC caps. The eXtreme/FV® Filter Vials are easy to press down with hands or toggle press.
These filter vials are manufactured with attention to detail and high-grade quality to ensure performance at optimal levels. Filter vials find a wide variety of applications ranging from life sciences, drug discovery, monitoring, and analysis. They are universal for all different types of autosamplers, including UHPLC.