ContactUsLinkComponent
NICHITIP UNIV F/RK/ST 100UL PK960
Nichitip Pipette Tips, Universal
# de catalogue 76808-752
Fournisseur:  Nichiryo America
NICHITIP UNIV F/RK/ST 100UL PK960
Nichitip Pipette Tips, Universal
# de catalogue 76808-752
Fournisseur:  Nichiryo America
Spécifications
  • Volume:
    2 - 100 µl
  • Filter:
    +
  • Sterile:
    +
  • Tip Style:
    Low retention
Spécifications
A propos de cet article
These tips are made of 100% virgin polypropyleylene.
  • High quality
  • Free of detectable DNase, RNase, DNA
  • Autoclavable
  • Low retention
Made in the highest quality moulds, these tips can be used whenever the work demands a higher standard.
Customers Who Bought This Also Bought