Premium Comfort Cleanroom Socks
# de catalogue 76795-312
Fournisseur: Vidaro
Spécifications
- Color:Blue
- Description:Premium comfort cleanroom socks
- Cat. no.:76795-312
Spécifications
A propos de cet article
Premium, form-fitting polyamide cleanroom sock.
- Heavy-duty polyamide cleanroom sock
- Provides thickness and breathability for ultimate comfort
- Flat toe seam for extra comfort
- One size fits all
Premium Comfort cleanroom socks feature a 97% polyamide/3% elastine construction that provides quality form-fitting comfort and breathability for wearers.
Customers Who Bought This Also Bought