CAP SNAP CLR PE 11MM RR/T PK100
# de catalogue TS6ARC11RRT1C
Fournisseur: Thermo Scientific
Customers Who Bought This Also Bought
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
194,14 $ to 694,12 $
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
635,20 $ to 2 541,50 $