CAP SCREW WHT PP 18MM CT F/T PK100
# de catalogue TS6ASC18-CTP
Fournisseur: Thermo Scientific
Customers Who Bought This Also Bought
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
635,20 $ to 2 541,50 $
Sodium hydroxide 40% (w/w) in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
146,85 $ to 576,51 $