ContactUsLinkComponent
MYPIPETMAN SELECT MULTI P12X10 0.5-10UL
MyPIPETMAN® Select Pipettors
# de catalogue 76716-738
Fournisseur:  GILSON, INC.
MYPIPETMAN SELECT MULTI P12X10 0.5-10UL
MyPIPETMAN® Select Pipettors
# de catalogue 76716-738
Fournisseur:  GILSON, INC.
Spécifications
  • Volume:
    0.5 - 10 µl
  • Description:
    P12×10
  • Compatible with:
    DL10, DFL10, DF30 and D200 diamond tips
  • Cat. no.:
    76716-738
Spécifications
A propos de cet article
Ergonomic, safe and reliable, MyPIPETMAN® Select offers volume setting choice and optional handle personalization for optimal use in various protocols.
  • Variable volume single channel models covering a volume range from 0.2 μl to 10 ml
  • Variable volume multichannel models covering a volume range from 0.5 μl to 300 μl (8 and 12-channel models)
  • Free volume setting position for quick and effortless volume adjustment
  • Light ratchet volume setting for precise volume adjustment
  • Volume locked position to secure pipetting
Set and secure pipetting volumes to meet specific needs and requirements. Simply adjust the volume by turning the push button of the pipette with the unique, patented Trilock™ volume-locking system. Lightweight and well-balanced, MyPIPETMAN® Select has an elegant and ergonomic design with a textured effect for a better grip. Without compromising on quality and performance, the built-in ejection system with an integrated lever provides reduced tip ejection forces compared to direct ejection systems. This patented ejection system reinforces the pipette’s ergonomics for convenient use throughout the day.
The MyPIPETMAN® Select offers optional handle printing on demand. Identify the pipette to avoid any risk of contamination by dedicating it to a specific liquid or workflow step and improve the lab's lean management process.
Customers Who Bought This Also Bought