Specifications
- Coating:Uncoated
- Description:Oiled
- Material:Stainless steel
- Thickness:0.004"
- Packed:250 blades/carton, 20 cartons/case
- Cat. no.:76717-294
- Blade Edge:3 facet
Specifications
About this item
AccuForge double edge blades feature a 3-Facet edge for exacting, clean cuts. Blades are individually wrapped for safety.
- 3-facet blade edge for increased sharpness
- High quality stainless steel
- Available with MicroCoat® enrichment technology
- Made in USA
AccuForge's MicroCoat® blade enrichment technology adds a layer of protection for each blade helping maintain a sharp effective edge over multiple uses.