ContactUsLinkComponent
BLADE DOUBLE EDGE SS UNCOAT OI CT250
AccuForge® Double Edge Blades, Individually Wrapped
Catalog # 76717-294
Supplier:  Accutec Blades
BLADE DOUBLE EDGE SS UNCOAT OI CT250
AccuForge® Double Edge Blades, Individually Wrapped
Catalog # 76717-294
Supplier:  Accutec Blades
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Coating:
    Uncoated
  • Description:
    Oiled
  • Material:
    Stainless steel
  • Thickness:
    0.004"
  • Packed:
    250 blades/carton, 20 cartons/case
  • Cat. no.:
    76717-294
  • Blade Edge:
    3 facet
Specifications
About this item
AccuForge double edge blades feature a 3-Facet edge for exacting, clean cuts. Blades are individually wrapped for safety.
  • 3-facet blade edge for increased sharpness
  • High quality stainless steel
  • Available with MicroCoat® enrichment technology
  • Made in USA
AccuForge's MicroCoat® blade enrichment technology adds a layer of protection for each blade helping maintain a sharp effective edge over multiple uses.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Customers Who Bought This Also Bought