Dropping Bottle
# de catalogue 470349-692
Fournisseur: United Scientific Supplies
Spécifications
- Capacity:60 ml
- Material:LDPE
- Cat. no.:470349-692
Spécifications
A propos de cet article
Dispense solutions accurately, one drop at a time.
- Ideal for storing and dispensing small volumes
- Translucent design
- LDPE with leakproof PP cap
- BPA-free
Dropping bottles are useful for adding small amounts of a liquid. With a small squeeze the amount of liquid being dispensed can be controlled. Manufactured from translucent and low density polyethylene. Caps are made from polypropylene. Assembled bottles are sold as packs.
Customers Who Bought This Also Bought