Medium Grade, 15 μm
# de catalogue 470349-724
Fournisseur: United Scientific Supplies
Spécifications
- Description:Filter Paper, Medium Grade, 70 mm diameter
- Cat. no.:470349-724
Spécifications
A propos de cet article
The most widely used qualitative filter paper for routine applications.
- Medium grade, 15 μm
Holds flocculants and precipitates commonly encountered in laboratory exercises.
Packaging: Packages of 100.
Customers Who Bought This Also Bought