Finntip™ Pipette Tips, Thermo Scientific
# de catalogue 76637-690
Fournisseur: Thermo Fisher Scientific
Spécifications
- Volume:1000 - 10000 µL
- Packed:5×24/Rack
- Sterile:+
- Tip Style:Finntip™ 10 mL
Spécifications
A propos de cet article
These high-quality pipette tips are specifically designed to fit Finnpipette® pipettors, but are also compatible with most pipettor brands.
- Made of high-quality polypropylene
- Autoclavable
- Chemical-resistant
Tips in refill trays can be used to quickly refill existing racks.
Customers Who Bought This Also Bought