ContactUsLinkComponent
BIOQUELL ISOLATOR CHEM INDICATORS CS100
Bioquell Isolator CI
Catalog # 76635-440
Supplier:  Eco Lab
BIOQUELL ISOLATOR CHEM INDICATORS CS100
Bioquell Isolator CI
Catalog # 76635-440
Supplier:  Eco Lab
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Isolator Chemical Indicator (Isolator CI)
  • Dimensions:
    Isolator CI
  • Size:
    100
  • Cat. no.:
    76635-440
Specifications
About this item
Bioquell offers different indicators to help you to verify bio-decontamination cycle completion and effectiveness. Chemical indicators (CI) can help you to verify a bio-decontamination cycle has achieved the expected vapor distribution and exposure. The Bioquell Isolator CI is for enclosures less than 10m3.
  • rapid-read chemical indicators for Ecolab Bioquell hydrogen peroxide vapor bio-decontamination for enclosed areas smaller than 10m3
  • Ideal for isolators, BSCs, filling lines, or small areas such as pass-throughs and MALs
  • Quickly confirm expected vapor distribution and exposure performance.
The Bioquell Isolator CIs are rapid-read chemical indicators for enclosed areas smaller than 10m^3. Use when bio-decontaminating isolators, BSCs, filling lines, or small areas such as pass-throughs and MALs, to quickly confirm expected vapor distribution and exposure performance.
Customers Who Bought This Also Bought