Specifications
- Description:Isolator Chemical Indicator (Isolator CI)
- Dimensions:Isolator CI
- Size:100
- Cat. no.:76635-440
Specifications
About this item
Bioquell offers different indicators to help you to verify bio-decontamination cycle completion and effectiveness. Chemical indicators (CI) can help you to verify a bio-decontamination cycle has achieved the expected vapor distribution and exposure. The Bioquell Isolator CI is for enclosures less than 10m3.
- rapid-read chemical indicators for Ecolab Bioquell hydrogen peroxide vapor bio-decontamination for enclosed areas smaller than 10m3
- Ideal for isolators, BSCs, filling lines, or small areas such as pass-throughs and MALs
- Quickly confirm expected vapor distribution and exposure performance.
The Bioquell Isolator CIs are rapid-read chemical indicators for enclosed areas smaller than 10m^3. Use when bio-decontaminating isolators, BSCs, filling lines, or small areas such as pass-throughs and MALs, to quickly confirm expected vapor distribution and exposure performance.