Order Entry
ContactUsLinkComponent
CAP 20MM BLUE CS1600
RayDyLyo® Sterile Snap Caps, DWK Life Sciences
# de catalogue 76632-786
RayDyLyo® Sterile Snap Caps, DWK Life Sciences
# de catalogue 76632-786

Spécifications

  • Color:
    Blue
  • Description:
    Snap cap with OmniFlex Igloo Lyo stopper
  • Size:
    20 mm
  • Cat. no.:
    76632-786

Spécifications

A propos de cet article

RayDyLyo® plastic push-fit caps.

  • Flip off snap caps are pre-assembled with an ISO 8362-2 compliant elastomer stopper
  • Container closure integrity (CCI) validated for use with ISO 8362-1 glass and COP vials
  • Delivered sterile (RTU) with a 2-year shelf life from time of manufacturer
  • Clean room manufactured with 100% in-process quality controls
  • Suitable for aseptic small batch, clinical trial, and robotic fill and finish operations
  • Snap caps do not require special crimping tools and affix with single vertical pressure
  • Provides tamper-evident closing and eliminates crimping defects
  • Compatible with gamma and steam terminal sterilization
  • Available in nested, standard bag, and RTP-type bag configurations

RayDyLyo® plastic push-fit caps from DWK Life Sciences, designed for closing vials of injectable drugs. These flip-off snap caps are pre-assembled with an ISO 8362-2 compliant elastomer stopper to simplify the production process.

Customers Who Bought This Also Bought