ContactUsLinkComponent
Value Plastics® Sanitary End Cap, 1¹/₂" Tri-Clamp, ADCF PP; 100/PK
Value Plastics® Sanitary End Cap, 1¹/₂" Tri-Clamp, ADCF PP; 100/PK
# de catalogue MFLX31513-18BLK
Fournisseur:  VALUE PLASTICS TE
Value Plastics® Sanitary End Cap, 1¹/₂" Tri-Clamp, ADCF PP; 100/PK
# de catalogue MFLX31513-18BLK
Fournisseur:  VALUE PLASTICS TE
  • Sterilisation:
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • En cours de visualisation
Spécifications
A propos de cet article
Certificate of compliance included!
  • Animal derivative component free
  • Meet USP Class IV requirements
  • Sterilizable vis gamma irradiation
These sanitary fittings are the perfect to transition from sanitary clamp connections to seal or barb for flexible tubing. All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow—and no particulate build-up. There are no sharp edges in the flow path, so cleaning is very easy. Made from animal-derived ingredient-free polypropylene, these adapters offer good chemical resistance and may be sterilized by gamma irradiation. Material meets the requirements for USP Class VI.
Delivery: Includes certificate of compliance.
  • Connection type:
    Sanitary Cap
  • Description:
    Sanitary End Cap, 1-1/2" Tri-Clamp, ADCF PP; 100/PK
  • Cat. no.:
    MFLX31513-18BLK
  • Fitting Material:
    Polypropylene
  • Flow Pattern:
    Straight
  • Sanitary Clamp Size (Port 1):
    1½"
  • Sanitary Gasket Size:
    1½"
Customers Who Bought This Also Bought