ContactUsLinkComponent
ROBOTIP HAM300C NF NS BL HT2 CS5760
J.T.Baker® Robotic Tips Type HT 2
Catalog # 49008-0015
   Low Manufacturing Impact
J.T.Baker® Robotic Tips Type HT 2
Catalog # 49008-0015
   Low Manufacturing Impact
Specifications
  • Volume:
    300 µl
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • Tip Color:
    Black
  • Packed:
    12× 5 Racks, 96 each, blistered
  • Cat. no.:
    49008-0015
  • Sterile:
    -
Specifications
About this item
Suitable for use on third party robotic instruments Hamilton Microlab Star and Nimbus. Available in conductive version, with or without filter.
  • Manufactured under cleanroom conditions (cleanroom Class 8)
  • State-of-the-art quality control
  • Ideal for high throughput analysis
  • Barcoded trays for automated identification
  • Packed in blister
J.T.Baker® robotic tips type HT 2 are suitable for third party robotic instruments Hamilton Microlab Star, Nimbus, and other robotic instruments using Hamilton’s CO-RE or CO-RE II pipetting head.
The validation of the tips on the instrument for specific applications is within the responsibility of the user.
Hamilton, Microlab Star™, Nimbus® and CO-RE® are registered trademarks of Hamilton company.
Customers Who Bought This Also Bought