Order Entry
ContactUsLinkComponent
PCR DNA LADDER 100-3000 BP 250LANES
VWR® DNA Ladders
# de catalogue 76620-560
Fournisseur:  VWR International
Numéro CAS:  
PCR DNA LADDER 100-3000 BP 250LANES
VWR® DNA Ladders
# de catalogue 76620-560
Fournisseur:  VWR International
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Description:
    PCR DNA Ladder, 100 to 3000 bp
  • Volume:
    1× 0.5 ml
  • Lanes:
    250 lanes (2 μl)

Spécifications

A propos de cet article

DNA ladders are supplied in a loading buffer that is ready to use on agarose and polyacrylamide DNA gels. The ladders are suitable with TBE, TAE, SB and LB electrophoresis systems.

  • Supplied in ready to use loading buffer
  • For direct loading and easy visualisation
  • Clear, distinct bands
  • Mass-calibrated bands for DNA quantification
  • PCR DNA LADDER has a 1000 bp band that is extra bright to serve as reference point
  • Blue dye front running at 100 to 300 bp at 0.5 to 1.5% agarose

High Range DNA Ladder molecular range: 200 to 12000 bp; mass-calibrated bands from 6 to 40 ng/band for DNA quantification.

Low Range DNA Ladder molecular range: 100 to 1000 bp; mass-calibrated bands from 8 to 40 ng/band for DNA quantification.

PCR DNA Ladder molecular range: 100 to 3000 bp; mass-calibrated bands from 10 to 30 ng/band for DNA quantification.

High Range DNA Ladder, Low Range DNA Ladder and PCR DNA Ladder are supplied in 0.5 mL packs sufficient for 250 lanes.

Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought